missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, Dihydrate, Purified, 95%, Spectrum™ Chemical

Catalog No. 18608990
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608990 500 g Poly Bottle
18608991 2.5 kg Poly Pail
18608992 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608990 Supplier Spectrum Chemical Mfg Cor Supplier No. B1027500GM
Only null left

Chemical Identifiers

CAS 10326-27-9

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95%
Grade Purified
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.