missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride dihydrate, Puratronic™, 99.997% (metals basis)

Catalog No. p-3256906
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1064614 25 g
AA1064622 100 g
AA1064636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1064614 Supplier Thermo Scientific Chemicals Supplier No. 01064614
Only null left

CAS: 10326-27-9 | BaCl2H4O2 | 244.26 g/mol

It is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Melting Point 113°C-2H2O
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
Formula Weight 244.28 (208.25 Anhydrous)
Odor Odorless
Appearance White powder
Density 3.097 g/cm3
Chemical Name or Material Barium chloride dihydrate, dihydrate
Show More Show Less
Hazard Category H301-H319-H332
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 600-412-6
RTECSNumber CQ8751000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only