missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| Color | White |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Physical Form | Crystals or Powder |
| Quantity | 100 g |
| Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
| Percent Purity | ≥99% |
| Grade | ACS |
| Loss on Drying | 14.0 to 16.0% |
| Packaging | White Poly Wide Mouth Bottle |
| Show More |
Safety and Handling
| ShelfLife | 60 months |