missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride dihydrate, 99+%

CAS: 10326-27-9 | BaCl2H4O2 | 244.26 g/mol

Supplier:  Thermo Scientific Chemicals A1290530

 View more versions of this product

Catalog No. AAA1290530


Only null left
Explore available promotions

Description

Description

Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.

Notes
Stable and hygroscopic in nature.
Specifications

Specifications

Barium chloride dihydrate
10326-27-9
BaCl2H4O2
UN1564
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
244.28 (208.25 Anhydrous)
Odorless
113°C (in 2H2O)
250 g
MFCD00149154
14,971
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
≥99%
3.097

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
O.O.[Cl-].[Cl-].[Ba++]
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c

H301-H319-H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

EINECSNumber : 233-788-1

RTECSNumber : CQ8751000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only