missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, Certified, 0.100N ±0.001N (0.05M), LabChem™
$167.00 - $357.93
Chemical Identifiers
| CAS | 10326-27-9 |
|---|---|
| Molecular Formula | BaCl2H4O2 |
| Molecular Weight (g/mol) | 244.26 |
| MDL Number | MFCD00149154 |
| InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
| Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
| PubChem CID | 5284346 |
| ChEBI | CHEBI:86153 |
| IUPAC Name | barium(2+) dihydrate dichloride |
| SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| 5284346 | |
| barium(2+) dihydrate dichloride |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| CHEBI:86153 | |
| O.O.[Cl-].[Cl-].[Ba++] |
Specifications
| 10326-27-9,7732-18-5 | |
| 98.78,1.22 | |
| 1 L | |
| BaCl2·2H2O | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+) dihydrate dichloride | |
| 5284346 | |
| 244.26 | |
| 0.100N ±0.001N (0.05M) | |
| Poly Bottle | |
| Barium Chloride |
| Colorless | |
| Liquid | |
| BaCl2H4O2 | |
| MFCD00149154 | |
| Soluble in water | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 | |
| CHEBI:86153 | |
| Certified | |
| Passes Test | |
| Hydrogen chloride; Barium |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature