missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride Anhydrous (Laboratory), Fisher Chemical™

Catalog No. p-31117
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
B31100C 100 g Poly Bottle
B31500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31100C Supplier Thermo Fisher Scientific Supplier No. B31 100C
Only null left

Chemical Identifiers

CAS 10361-37-2
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+) dichloride
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560°C
Melting Point 960°C
Color White
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥97%
UN Number UN 1564
Formula Weight 208.23
Percent Purity ≥97%
Grade Laboratory
Identification Pass Test
Packaging Poly Bottle
Density 3.86g/cm³
Chemical Name or Material Barium Chloride Anhydrous
Show More Show Less
DOTInformation DOT Class 6.1, : Poison