missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, Anhydrous, 98%, Spectrum™ Chemical

Catalog No. p-7165125
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608981 125 g Poly Bottle
18608983 500 g Poly Bottle
18608984 2.5 kg Poly Pail
18608982 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608981 Supplier Spectrum Chemical Mfg Cor Supplier No. B1028125GM
Only null left

Chemical Identifiers

CAS 10361-37-2

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.