missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, ACS Grade, ≥ 99.0%, LabChem™

Supplier:  LabChem LC115601

 View more versions of this product

Catalog No. LC115601


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1560°C
963°C
White
Crystals
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
244.26
ACS
Poly Bottle
Hydrogen chloride; Barium
Barium Chloride
10326-27-9
100
500 g
BaCl2·2H2O
UN1564
Soluble in water
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
≥99.0%
Passes Test
3100kg/m3
3100kg/m3

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Harmful to aquatic life.

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions