missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, ACS Grade, ≥ 99.0%, LabChem™

Catalog No. LC115609
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC115609 100 g
LC115601 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC115609 Supplier LabChem Supplier No. LC115609
Only null left

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560°C
Melting Point 963°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 100 g
Linear Formula BaCl2·2H2O
UN Number UN1564
Solubility Information Soluble in water
Formula Weight 244.26
Percent Purity ≥99.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 3100kg/m3
Decomposition Information Hydrogen chloride; Barium
Density 3100kg/m3
Chemical Name or Material Barium Chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature