Learn More
Barium chloranilate trihydrate
CAS: 32458-20-1 | C6H8BaCl2O7 | 400.35 g/mol
$406.95 - $1320.79
Chemical Identifiers
| CAS | 32458-20-1 |
|---|---|
| Molecular Formula | C6H8BaCl2O7 |
| Molecular Weight (g/mol) | 400.35 |
| MDL Number | MFCD00078327 |
| InChI Key | JFBLKYVZQCFPJE-UHFFFAOYSA-N |
| Synonym | barium chloranilate, reag |
| PubChem CID | 131877766 |
| IUPAC Name | barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate |
| SMILES | O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC401660250
|
Thermo Scientific Chemicals
401660250 |
25 g | Plastic bottle |
Each for $406.95
|
|
||||
|
AC401661000
|
Thermo Scientific Chemicals
401661000 |
100 g | Plastic bottle |
Each for $1,320.79
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 32458-20-1 | |
| 400.35 | |
| JFBLKYVZQCFPJE-UHFFFAOYSA-N | |
| 131877766 | |
| O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O |
| C6H8BaCl2O7 | |
| MFCD00078327 | |
| barium chloranilate, reag | |
| barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate |
Specifications
| 32458-20-1 | |
| Authentic | |
| C6H8BaCl2O7 | |
| 25 g | |
| Solubility in water: soluble | |
| O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O | |
| 400.35 | |
| 398.34 | |
| Barium chloranilate trihydrate |
| Brown | |
| Plastic bottle | |
| MFCD00078327 | |
| barium chloranilate, reag | |
| JFBLKYVZQCFPJE-UHFFFAOYSA-N | |
| barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate | |
| 131877766 | |
| Powder |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 236-567-8
TSCA : TSCA
RUO – Research Use Only