missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium bromide, 99%, pure, anhydrous, Thermo Scientific™
For ultra trace analysis
Chemical Identifiers
| CAS | 10553-31-8 |
|---|---|
| Molecular Formula | BaBr2 |
| Molecular Weight (g/mol) | 297.16 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC315380500
|
Thermo Scientific Chemicals
315380500 |
50g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC315382500
|
Thermo Scientific Chemicals
315382500 |
250g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 10553-31-8 | |
| BaBr2 | |
| 297.16 |
Specifications
| 10553-31-8 | |
| 98.5% min. (Argentometry) | |
| BaBr2 | |
| Solubility in water: 104.1g/100mL (20°C). Other solubilities: soluble in methanol, almost insoluble in ethanol, ethyl acetate,, acetone, dioxane | |
| 297.16 | |
| Pure |
| 50g | |
| BaBr2 | |
| 15,964 | |
| 297.16 | |
| 99% | |
| Barium bromide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Warning
EINECSNumber : 234-140-
RUO â Research Use Only