missing translation for 'onlineSavingsMsg'
Learn More

Barium bromate, 97%

Catalog No. AAA1100822
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1100822 100 g
AAA1100836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1100822 Supplier Thermo Scientific Chemicals Supplier No. A1100822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13967-90-3 | BaBr2O6 | 393.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13967-90-3
Molecular Formula BaBr2O6
Molecular Weight (g/mol) 393.129
MDL Number MFCD00014177
InChI Key VEASZGAADGZARC-UHFFFAOYSA-L
Synonym barium bromate, bromic acid, barium salt, unii-5qk4064y0r, barium 2+ dibromate, acmc-1bumj, ba.2bro3, bromic acid, barium salt 2:1, barium bromate un2719 oxidizer
PubChem CID 61706
IUPAC Name barium(2+);dibromate
SMILES [O-]Br(=O)=O.[O-]Br(=O)=O.[Ba+2]

Specifications

Quantity 100 g
Odor Odorless
Density 3.99
UN Number UN2719
Formula Weight 393.16
Percent Purity 97%
Chemical Name or Material Barium bromate
Hazard Category H272-H302+H332
Hazard Statement GHS H Statement
H272-H302-H332
May intensify fire
oxidizer.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM BROMATE
EINECSNumber 237-750-5
RTECSNumber EF8715000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only