Learn More
Barium aluminum oxide, tech.
CAS: 12004-04-5
$35.30 - $434.26
Chemical Identifiers
| CAS | 12004-04-5 |
|---|---|
| MDL Number | MFCD00049716 |
| Synonym | Barium aluminate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA2264209
|
Thermo Scientific Chemicals
2264209 |
10 g |
Each for $35.30
|
|
|||||
|
AA2264222
|
Thermo Scientific Chemicals
02264222 |
100 g |
Each for $133.45
|
|
|||||
|
AA2264236
|
Thermo Scientific Chemicals
02264236 |
500 g |
Each for $434.26
|
|
|||||
Description
Barium aluminum oxide is used for getter alloys in vacuum tubes. It also finds application as deoxidizer for copper, Fraryâs metal, lubricant for anode rotors in x-ray tubes and spark-plug alloys. It is used to prepare heat-resistant refractory and super-refractory concretes. Further, it gives a better protection against gamma radiation and X-rays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12004-04-5 | |
| MFCD00049716 | |
| Barium aluminate |
Specifications
| 12004-04-5 | |
| 10 g | |
| UN1564 | |
| Insoluble in water. | |
| Technical | |
| Barium aluminum oxide |
| Powder | |
| MFCD00049716 | |
| Barium aluminate | |
| 561.98 | |
| Odorless |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber : 234-445-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only