missing translation for 'onlineSavingsMsg'
Learn More

Barium Acrylate Monomer 95.0+%, TCI America™

Catalog No. A076025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A076025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A076025G Supplier TCI America Supplier No. A076025G
Only null left

Chemical Identifiers

CAS 17989-90-1
Molecular Formula C6H6BaO4
Molecular Weight (g/mol) 279.437
MDL Number MFCD00067204
InChI Key ONQPQIFFWHMGGE-UHFFFAOYSA-L
Synonym Acrylic Acid Barium Salt Monomer
PubChem CID 13609154
IUPAC Name barium(2+);prop-2-enoate
SMILES C=CC(=O)[O-].C=CC(=O)[O-].[Ba+2]

Specifications

Color White
Quantity 25 g
UN Number 1564
Formula Weight 279.44
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Barium Acrylate Monomer