missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, Puriss. p.a., ACS Reagent, ≥99%, Solstice

Catalog No. 6002415
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002415 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002415 Supplier Solstice Supplier No. 323051KG

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Physical Form Solid
Quantity 1 kg
Packaging Fiberboard Box
Linear Formula (CH3COO)2Ba
UN Number NONH for all modes of transport
Beilstein 3693411
IUPAC Name barium(2+);diacetate
Formula Weight 255.42g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Barium Acetate
Show More Show Less
Hazard Statement P261-P301 + P312 + P330
Precautionary Statement H302 + H332
EINECSNumber 208-849-0