missing translation for 'onlineSavingsMsg'
Learn More

Barium Acetate, Granular, ACS, 99.0-102.0%, Spectrum™ Chemical

Catalog No. p-7165119
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608958 125 g Poly Bottle
18608959 500 g Poly Bottle
18608957 1 kg Poly Bottle
18608962 2.5 kg Poly Pail
18608960 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608958 Supplier Spectrum Chemical Mfg Cor Supplier No. B1010125GM
Only null left

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
IUPAC Name barium(2+) diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99 to 102%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.