missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, ACS, 99.0-102.0%

Catalog No. p-7048956
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1219830 250 g
AA12198A1 1 kg
AA12198A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1219830 Supplier Thermo Scientific Chemicals Supplier No. 01219830
Only null left

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol

Barium acetate is used as a chemical reagent, textile mordant and lubricant. It is used to prepare other acetates and barium carbonate. It acts as a mordant for printing textile fabrics. It is utilized for drying paints and varnishes. It serves as a catalyst in organic synthesis. It plays an important role in the separation of glycosaminoglycans. Further, it acts as a cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. In addition, it is used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+);diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline
Quantity 250 g
UN Number UN1564
Merck Index 14,965
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 255.43
Percent Purity 99.0 to 102.0%
Grade ACS
Odor Odorless
Density 2.47 g/mL
Chemical Name or Material Barium acetate
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 208-849-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only