missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, 99%

Catalog No. AAA1031630
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1031630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1031630 Supplier Thermo Scientific Chemicals Supplier No. A1031630

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol

It has acted as cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. Barium acetate has been used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions. Electrophoresis in the presence of barium acetate has analytically and preparatively separate glycosaminoglycans. It has been documented to induce severe cardiac disrhythmia from ingestion. It also finds its application as a chemical reagent, textile mordant, catalyst for organic reactions, and paint and varnish driers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has acted as cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. Barium acetate has been used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions. Electrophoresis in the presence of barium acetate has analytically and preparatively separate glycosaminoglycans. It has been documented to induce severe cardiac disrhythmia from ingestion. It also finds its application as a chemical reagent, textile mordant, catalyst for organic reactions, and paint and varnish driers.

Solubility
Soluble in water at 20°C 588g/L. Insoluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications

CAS 543-80-6
Density 2.47
Odor Odorless
Molecular Formula C4H6BaO4
Quantity 250 g
MDL Number MFCD00012447
UN Number UN1564
Merck Index 14,965
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
Solubility Information Soluble in water at 20°C 588g/L. Insoluble in alcohol.
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES [Ba++].CC([O-])=O.CC([O-])=O
IUPAC Name barium(2+);diacetate
Molecular Weight (g/mol) 255.42
PubChem CID 10980
Formula Weight 255.43
Percent Purity 99%
Chemical Name or Material Barium acetate
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 208-849-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only