missing translation for 'onlineSavingsMsg'
Learn More

Barium, 99+%, rods, approx. 2 cm diameter

CAS: 7440-39-3 | Ba | 137.33 g/mol

$501.11 - $2996.34

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC317861000
SDS View Documents
Thermo Scientific Chemicals
317861000
100 g
Each for $501.11
Only null left
 
AC317865000
SDS View Documents
Thermo Scientific Chemicals
317865000
500 g
Each for $2,996.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-39-3
137.33
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
5355457
barium
Ba
MFCD00134031
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
CHEBI:32595
[Ba]

Specifications

Gray
7440-39-3
99.0
100 g
Rod
Ba
MFCD00134031
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
[Ba]
137.33
CHEBI:32595
≥99%
725.0°C
1640.0°C
100.0
Plastic bottle
99+%
Ba
15, 960
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
barium
5355457
137.34
Barium, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
In contact with water releases flammable gas.
Reacts violently with water.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only