missing translation for 'onlineSavingsMsg'
Learn More

Barium 2,4-pentanedionate

CAS: 12084-29-6 | C10H14BaO4 | 335.55 g/mol

$192.42 - $192.42

Chemical Identifiers

CAS 12084-29-6
Molecular Formula C10H14BaO4
Molecular Weight (g/mol) 335.55
MDL Number MFCD00013484
InChI Key DJHZYHWLGNJISM-FDGPNNRMSA-L
Synonym barium 2+ ; z-4-oxopent-2-en-2-olate, barium 2,4-pentanedionate, bis pentane-2,4-dionato-o,o' barium, barium, bis 2,4-pentanedionato-o,o'-, t-4, 2,4-pentanedione,ion 1-, barium 2:1, barium, bis 2,4-pentanedionato-o,o'-, hydrate
PubChem CID 5486157
SMILES [Ba++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1461322
View Documents
Thermo Scientific Chemicals
01461322
100 g
Each for $192.42
Only null left
 
Description

Description

Barium 2,4-pentanedionate hydrate is used in the preparation of radioluminescence of poly ethylene glycolated Eu3+ doped nanophosphors as bioimaging probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12084-29-6
335.55
DJHZYHWLGNJISM-FDGPNNRMSA-L
5486157
C10H14BaO4
MFCD00013484
barium 2+ ; z-4-oxopent-2-en-2-olate, barium 2,4-pentanedionate, bis pentane-2,4-dionato-o,o' barium, barium, bis 2,4-pentanedionato-o,o'-, t-4, 2,4-pentanedione,ion 1-, barium 2:1, barium, bis 2,4-pentanedionato-o,o'-, hydrate
[Ba++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

>320°C
Powder
C10H14BaO4
UN1564
Insoluble in water.
[Ba++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
335.55
335.56 (Anhydrous)
12084-29-6
100 g
MFCD00013484
barium 2+ ; z-4-oxopent-2-en-2-olate, barium 2,4-pentanedionate, bis pentane-2,4-dionato-o,o' barium, barium, bis 2,4-pentanedionato-o,o'-, t-4, 2,4-pentanedione,ion 1-, barium 2:1, barium, bis 2,4-pentanedionato-o,o'-, hydrate
DJHZYHWLGNJISM-FDGPNNRMSA-L
barium(2+);(Z)-4-oxopent-2-en-2-olate
5486157
Barium 2,4-pentanedionate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

EINECSNumber : 235-151-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only