missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Bare Copper Wire

This Bare Copper Wire is measured by mass.

Supplier:  Eisco™ PH1058G

 View more versions of this product

Catalog No. S24905


Only null left

Description

Description

  • Available as 50 ft. reel and 500 g roll.
  • Includes one spool.
Specifications

Chemical Identifiers

Copper
Bare Wire
1 Ea.

Specifications

Copper
0.018 in.
1 Ea.
0.46 mm
Bare Wire
167.6 m
550 ft.
26 SWG (24/25 AWG)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions