missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Bare Copper Wire

This Bare Copper Wire is measured by mass.

Supplier:  Eisco™ PH1058F

 View more versions of this product

Catalog No. S24904


Only null left

Description

Description

  • Available as 50 ft. reel and 500 g roll.
  • Includes one spool.
Specifications

Chemical Identifiers

Copper
Bare Wire
1 Ea.

Specifications

114.3 m
1 Ea.
Bare Wire
375 ft.
0.022 in.
Copper
24 SWG (23/24 AWG)
0.56 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions