missing translation for 'onlineSavingsMsg'
Learn More

Aspen Surgical™ Bard-Parker™ Sterile Scalpels

Intended for tissue separation, and other procedures that require a sharp surgical blade to puncture or cut

$68.97 - $68.97

Specifications

Blade Material Stainless Steel
Product Type Surgical Scalpel
For Use With (Application) Tissue Separation and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut
Disposable Single-use
Sterility Sterile
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
14-823-45
View Documents
Aspen Surgical™
371610
# 10 N/A
Only null left
 
Description

Description

  • Offers the superior cutting advantages of stainless steel technology
  • Consistent sharpness, control and strength surgeons demand
  • Uniquely packaged for safety and ease of use
  • Sterile
  • Not made with natural rubber latex
Specifications

Specifications

Stainless Steel
Tissue Separation and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut
Sterile
Surgical Scalpel
Single-use
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'