Learn More
StatLab Bard-Parker Scalpel Blade
Bard-Parker Scalpel Blade
Supplier: StatLab BP1322
Description
- Bard-Parker rib-back scalpel blades offer the superior cutting advantages of carbon steel technology
- The exclusive Rib-Back blade design for greater strength and control
- Specially ground and carefully inspected
- Packaged 150 per box, non-sterile and individually wrapped
Chemical Identifiers
| Blade | |
| Non-sterile | |
| No |
| Single-use | |
| Not Autoclavable |
Specifications
| Blade | |
| Single-use | |
| Not Autoclavable |
| # 22 | |
| Non-sterile | |
| No |