Learn More
StatLab Bard-Parker Scalpel Blade
Catalog No. 22000019
Change view
Click to view available options
Size:
# 22
# 60
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22000019 # 60
22000018 # 22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000019 Supplier StatLab Supplier No. BP1340
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bard-Parker Scalpel Blade
  • Bard-Parker rib-back scalpel blades offer the superior cutting advantages of carbon steel technology
  • The exclusive Rib-Back blade design for greater strength and control
  • Specially ground and carefully inspected
  • Packaged 150 per box, non-sterile and individually wrapped
Specifications
Product Type Blade
Size # 60
Disposable Single-use
Sterility Non-sterile
Autoclavable Not Autoclavable
Graduated No