missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspen Surgical™ Bard-Parker™ Protected Disposable Scalpel
Features one-handed, locking retractable shield
$127.04
Specifications
| Blade Material | Stainless Steel |
|---|---|
| Product Type | Safety Scalpel |
| For Use With (Application) | Tissue Separation and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut |
| Disposable | Single-use |
| Sterility | Sterile |
Description
Features one-handed, locking retractable shield
- Designed to minimize the risk of surgical blade injuries
- Features a BD Bard-Parker stainless-steel blade, nonslip grip, and a centimeter scale on the plastic, surgical-green handle
- Clear protective shield locks both when the blade is covered for protection before and after use, and when blade is uncovered for security and control during use
- Top button provides tactile sensation as well as an audible click to confirm lock; allows for left or right hand activation
- Indicator line offers visual confirmation of lock
- Autoclavable for procedural kits and on-site sterilization
Specifications
| Stainless Steel | |
| Tissue Separation and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut | |
| Sterile | |
| Autoclavable |
| Safety Scalpel | |
| Single-use | |
| OSHA |