Learn More
Aspen Surgical™ Bard-Parker™ Carbon Steel Blade with Rib-Back™ Design
Surgical carbon steel, ribbed for extra rigidity
Supplier: Aspen Surgical™ 371340
Description
Surgical carbon steel, ribbed for extra rigidity
- Convex cutting edge
- 2.37 in. long (6cm)
- Nonsterile
- Fits No. 8 handle only
- Bard-Parker™ brand
- Individually wrapped
Specifications
| Dissecting Blade | |
| # 60 | |
| 6 cm | |
| Non-sterile |
| Ribbed | |
| 2.37 in. | |
| For Dissecting Knife Handles |