Learn More
StatLab Bard-Parker Blade Handle
Catalog No. 22000017
Change view
Click to view available options
Size:
No. 6
No. 8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22000017 No. 8
22000016 No. 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000017 Supplier StatLab Supplier No. STLBP1080
Only null left
Bard-Parker Blade Handle
  • Intended to be used with Bard-Parker conventional blades - BP1322 and BP1340
  • High-impact poly handles are designed for a contoured, firm grip and easy manipulation
Specifications
Product Type Detachable Handle
Size No. 8
Disposable Reusable
Sterility Non-sterile
Autoclavable Not Autoclavable
Graduated No