Learn More
StatLab Bard-Parker Blade Handle
Description
- Intended to be used with Bard-Parker conventional blades - BP1322 and BP1340
- High-impact poly handles are designed for a contoured, firm grip and easy manipulation
Specifications
Specifications
| Product Type | Detachable Handle |
| Size | No. 6 |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Graduated | No |