missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barcoded Latch Racks for Matrix™ Storage Tubes
GREENER_CHOICE

Catalog No. 04100559
Change view
Click to view available options
For Use With (Equipment):
0.75mL tubes
1.0mL Screwtop Tubes
1.4mL tubes
500μL Screwtop Tubes
500μL Tubes
Quantity:
10 Case
5 Case
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
04100559 1.4mL tubes 10 Case
14754691 1.0mL Screwtop Tubes 5 Case
14754693 500μL Tubes 10 Case
14754696 500μL Screwtop Tubes 5 Case
04100560 0.75mL tubes 10 Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 04100559 Supplier Thermo Scientific™ Supplier No. 4902

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Create custom arrays with empty ANSI-format racks featuring 2D and linear barcodes and human readable type on three sides and a 2D barcode on the base of the rack.

Enhance sample storage at low temperatures, including vapor phase Liquid Nitrogen, with Thermo Scientific™ Matrix™ barcoded latch racks. Barcoded Latch Racks feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack for the ultimate in sample tracking. Use in combination with Thermo Scientific 2D barcoded tubes, barcode readers, arrayers and capping equipment.

Superior Storage Rack Design

  • Storage in robot-friendly ANSI-footprint rack
  • Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking regardless of your barcode system
  • Rack barcodes are compatible with our range of VisionMate readers
  • Hinged lid rests in “open” position on rack while conducting manual pipetting to prevent contamination of lid sitting on bench

Latch Racks Save Precious Space

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Latch rack lid can be lifted robotically for tube access with high-throughput automated applications

Specifications

Description 1.4mL Latch Rack Lids
Capacity (Metric) 1.4 mL
For Use With (Equipment) 1.4mL tubes
Quantity 10 Case
Capacity (English) 0.0473 fl oz.
Product Type Barcoded Latch Rack