missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Bar-coded Cryogenic Vials

Catalog No. 07200963
Change view
Click to view available options
Barcode:
1D
2D
Thread Style:
External
External threaded
Internal
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Barcode Thread Style
07200963 1D External
07200960 2D Internal
07200961 2D External threaded
07200962 1D Internal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07200963 Supplier Corning™ Supplier No. 8676
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For storing biological and compound samples in labeled vials at cryogenic temperatures. Corning™ Bar-coded Cryogenic Vials provides a validated unique bar-code that will not fade or fall off.

  • Manufactured from polypropylene
  • Leak proof seal for integrity of storage
  • Autoclavable vials and caps, can be re-sterilized
  • Unique 1D, 2D bar-codes and human readable characters
  • Features star shaped cavity in cap and is compatible with decapping equipment
  • Locking base stabilizes tubes for automated handling
  • Graduations and writing patch for manual use also
  • Each tube is checked for clarity and readability of 2D code

Specifications

Closure Color Orange
Color Natural
Sterility Sterile
Volume (Metric) 2 mL
For Use With (Application) Cell biology, cell banking, compound cryopreservation
No. per Case 500
No. per Pack 50
Thread Style External
Base Style Round, Self-standing
Barcode 1D
Closure Material Polyethylene
Show More Show Less