missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences BarbLock™ Tubing Retainer, PVDF

BarbLock™ ultra-secure retainers are the most advanced design available in today's demanding connective applications. BarbLock™ retainers are the only system with a full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge.

$1,996.07 - $2,836.57

Specifications

Certifications/Compliance USP Class VI
Autoclavable Not Autoclavable
Material PVDF
For Use With Flexible tubing, Compatible with Pure-Fit SIB™ fittings
Packaging Quantity 250/Bag
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
13417573
Encompass
View Documents Promotion Details
Saint Gobain Life Sciences
BL135125ALPVDF
125 Series, AL (One-piece)
Pack of 2 Each for $1,996.07
Only null left
 
13417582
Encompass
View Documents Promotion Details
Saint Gobain Life Sciences
BL135252ALPVDF
252 Series, AL (One-piece)
Pack of 4 Each for $2,836.57
Only null left
 
Description

Description

When it comes to retaining flexible tubing and hose onto barbed fittings, BarbLock™ retainers have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. The retainer uses no adhesives or solvents, is ideal for systems transferring delicate liquids or gases, and is quick and easy to assemble, saving valuable time. The standard retainer is a two-piece unit (sleeve and collet), while the one-piece AL retainer combines the sleeve and collet and can be automatically loaded into assembly.

  • Replaces inconsistent and leak-prone cable ties and traditional clamps
  • Compatible with all flexible tubing materials
  • Provides 360° radial crimp behind the barb
  • Supplies 360° compression over the barb and full dynamic retention
  • Eliminates solvent bonding, ultraviolet bonding and toxic adhesives
  • Constructed with high-performance PVDF resin that is USP Class VI compliant
  • Quick and easy to assemble
  • Sterilize by gamma irradiation
  • Color: Blue

Order Info

13-417-539 and 13-417-544 come as pack of one.

Specifications

Specifications

USP Class VI
PVDF
250/Bag
Blue
Not Autoclavable
Flexible tubing, Compatible with Pure-Fit SIB™ fittings
Gamma
Tubing Connector
Videos
SDS
Documents

Documents