missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain BarbLock™ Tubing Retainer, Polypropylene

BarbLock™ ultra-secure retainers are the most advanced design available in today's demanding connective applications. BarbLock™ retainers are the only system with a full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge.

Specifications

Certifications/Compliance USP Class VI
Autoclavable Not Autoclavable
Material Polypropylene
For Use With Flexible tubing, Compatible with Pure-Fit SIB™ fittings
Color Blue
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
13417544
Encompass
View Documents Promotion Details
Saint-Gobain
BL135250
250 Series, Standard N/A
Only null left
 
Description

Description

When it comes to retaining flexible tubing and hose onto barbed fittings, BarbLock™ retainers have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. The retainer uses no adhesives or solvents, is ideal for systems transferring delicate liquids or gases, and is quick and easy to assemble, saving valuable time. The standard retainer is a two-piece unit (sleeve and collet), while the one-piece AL retainer combines the sleeve and collet and can be automatically loaded into assembly.

  • Replaces inconsistent and leak-prone cable ties and traditional clamps
  • Compatible with all flexible tubing materials
  • Provides 360° radial crimp behind the barb
  • Supplies 360° compression over the barb and full dynamic retention
  • Eliminates solvent bonding, ultraviolet bonding and toxic adhesives
  • Constructed with high-performance polypropylene resin that is USP Class VI compliant
  • Quick and easy to assemble
Specifications

Specifications

USP Class VI
Polypropylene
Blue
Not Autoclavable
Flexible tubing, Compatible with Pure-Fit SIB™ fittings
Videos
SDS
Documents

Documents