missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain BarbLock™ Tubing Retainer, Polypropylene

Catalog No. 13417544
Encompass
Change view
Click to view available options
Product Type:
250 Series, Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
13417544 250 Series, Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13417544 Supplier Saint-Gobain Supplier No. BL135250

BarbLock™ ultra-secure retainers are the most advanced design available in today's demanding connective applications. BarbLock™ retainers are the only system with a full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge.

When it comes to retaining flexible tubing and hose onto barbed fittings, BarbLock™ retainers have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. The retainer uses no adhesives or solvents, is ideal for systems transferring delicate liquids or gases, and is quick and easy to assemble, saving valuable time. The standard retainer is a two-piece unit (sleeve and collet), while the one-piece AL retainer combines the sleeve and collet and can be automatically loaded into assembly.

  • Replaces inconsistent and leak-prone cable ties and traditional clamps
  • Compatible with all flexible tubing materials
  • Provides 360° radial crimp behind the barb
  • Supplies 360° compression over the barb and full dynamic retention
  • Eliminates solvent bonding, ultraviolet bonding and toxic adhesives
  • Constructed with high-performance polypropylene resin that is USP Class VI compliant
  • Quick and easy to assemble

Specifications

Certifications/Compliance USP Class VI
Autoclavable Not Autoclavable
Material Polypropylene
For Use With Flexible tubing, Compatible with Pure-Fit SIB™ fittings
Packaging Quantity 250/Bag
Sterilization Method Gamma
Color Blue
Product Type 250 Series, Standard