missing translation for 'onlineSavingsMsg'
Learn More

Barbituric Acid, 99%, Spectrum™ Chemical

Catalog No. 18608956
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
Packaging:
2500cc White opaque plastic container
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608956 500 g Poly Bottle
18608954 25 g Poly Bottle
18608952 125 g Poly Bottle
18608953 2.5 kg 2500cc White opaque plastic container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608956 Supplier Spectrum Chemical Mfg Cor Supplier No. BA120500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-52-7

Specifications

CAS Max % 1
Percent Purity 99%
Grade Reagent
Quantity 500 g
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.