missing translation for 'onlineSavingsMsg'
Learn More

Barbituric Acid, 98% (Malonylurea; Pyrimidinetrione), Reagents

Catalog No. 01337475
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337475 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337475 Supplier Labchem Inc Supplier No. C2145500100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Barbituric Acid, 98% (Malonylurea; Pyrimidinetrione), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-52-7
Molecular Formula C4H4N2O3
Molecular Weight (g/mol) 128.09
InChI Key HNYOPLTXPVRDBG-UHFFFAOYSA-N
Synonym 1,3-Diazinane-2,4,6-trione, Malonylurea, 2,4,6-Pyrimidinetrione
IUPAC Name 1,3-diazinane-2,4,6-trione
SMILES O=C1CC(=O)NC(=O)N1

Specifications

Melting Point 248°C
Boiling Point 260°C
Color White to Off-White
Quantity 100 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 1
Grade Reagent
Chemical Name or Material Barbituric Acid, 98% (Malonylurea, Pyrimidinetrione)