missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Bar Breaking Apparatus

Catalog No. S23743
Change view
Click to view available options
Grade:
7 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S23743 7 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23743 Supplier Eisco™ Supplier No. PH0380A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use the Bar Breaking Apparatus to demonstrate the forces which can be exerted during thermal expansion or contraction.

  • Heavy cast iron frame with slotted end pillars to carry stout iron bar.
  • Bar threaded at one end for large tensioning nut and holes at other end to accommodate cast iron breaking bars.
  • Supplied with 10 breaking bars.

Specifications

Grade 7 to 12
For Use With (Application) Demonstrate the forces which can be exerted during thermal expansion or contraction
Includes 10 breaking bars
Material Cast Iron
Dimensions (L x W x H) 16.9 x 4.3 x 4.3 in.
Product Type Bar Breaking Apparatus