missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BAPTA, Tetra sodium Salt, Calbiochem™,

Catalog No. 1964181gm
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1964181GM 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1964181GM Supplier MilliporeSigma™ Supplier No. 1964181GM

Chemical Identifiers

CAS 126824-24-6
Molecular Formula C22H20N2Na4O10
Molecular Weight (g/mol) 564.37
MDL Number MFCD02683087
InChI Key ZWSMLJACYSGFKD-UHFFFAOYSA-J
Synonym bapta-na4, bapta tetrasodium, bapta tetrasodium salt, sodium 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, bapta, tetrasodium salt, bapta tetrasodium salt hydrate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrasodium salt, tetrasodium 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetate, 1,2-bis 2-aminophenoxy-ethane-n,n,n, n-tetraacetic acid tetrasodium salt, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrasodium salt hplc
PubChem CID 2734965
IUPAC Name tetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC([O-])=O)CC([O-])=O

Specifications

Color White
Packaging Glass Bottle
Quantity 1 g
Physical Form Solid