missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bandmate™ Reagent Tubes
Description
The tubes are required for dispensing primary and secondary antibodies into the Bandmate mini and midi incubation trays during the automated western blot processing procedure. Two reagent tubes are required for each blot.
Specifications
Specifications
| Color | Clear |
| Description | Bandmate Reagent Tubes, 500 Pack |
| For Use With (Application) | Western Blot |
| For Use With (Equipment) | Invitrogen Bandmate Automated Western Blot Processor |
| Product Line | Bandmate |
| Type | Reagent Tube |
| Quantity | 500 tubes |
| Shipping Condition | Ambient |