missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bandmate™ Blot Tray Cover

Catalog No. BW0050
Encompass
Change view
Click to view available options
Quantity:
1 Tray Cover
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW0050 1 Tray Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BW0050 Supplier Invitrogen™ Supplier No. BW0050
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Bandmate Blot Tray Cover is a replacement cover specially designed for use with the Bandmate Automated Western Blot Processor mini and midi incubation trays

This tray cover is required to protect blots and reagents during western blot processing.

Specifications

Color Clear
Description Bandmate Tray Cover
For Use With (Application) Western Blot
For Use With (Equipment) Invitrogen Bandmate Automated Western Blot Processor
Product Line Bandmate
Type Tray Cover
Quantity 1 Tray Cover
Shipping Condition Ambient
Dimensions (L x W x H) 21 cm x 12.7 cm x 0.953 cm
Weight 0.091 kg
Weight (English) 3.2 oz
Weight (Metric) 0.091 kg
Show More Show Less