missing translation for 'onlineSavingsMsg'
Learn More

Allegro™ Banana Oil Fit-Test Solutions

Catalog No. 19088507
Change view
Click to view available options
Capacity (English):
1 oz.
Capacity (Metric):
29.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Capacity (Metric)
19088507 1 oz. 29.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19088507 Supplier Allegro™ Supplier No. 020301

Isoamyl Acetate, also referred to as Banana Oil, is a qualitative fit test agent used to detect respirator leakage.

  • Banana oil (isoamyl acetate) is the substance originally used for testing respirator fit; its strong, recognizable odor quickly indicates the lack of proper fit
  • Available in two fit test kits and Fit Check Ampules that provide a fast and easy method of on-the-spot qualitative fit testing

Order Info

The Standard Kit comes in a handy carry bag and includes two 1 oz. (30mL) bottles of testing agent, a vinyl test enclosure, measuring pipette, and paper towels. The Deluxe Kit comes in a convenient storage case and contains everything necessary to perform a proper fit test, including an 8 1oz. (30mL) bottles of testing solution, a fit test tent, jars, paper towels, disposable bags, and other necessary components. Replacement banana oil solution is available in 1 oz. (30mL) bottles.

Specifications

Product Type Banana Oil Solution
Capacity (English) 1 oz.
Capacity (Metric) 29.5 mL
For Use With (Application) Respirator Fit Testing
Model 0203-01
Quantity 1 Ea.