missing translation for 'onlineSavingsMsg'
Learn More

Balsalazide Disodium Salt Dihydrate 98.0+%, TCI America™

Catalog No. p-7123362
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41211G 1 g
B41215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41211G Supplier TCI America Supplier No. B41211G
Only null left

Chemical Identifiers

CAS 150399-21-6
Molecular Formula C17H17N3Na2O8
Molecular Weight (g/mol) 437.316
MDL Number MFCD01631117
InChI Key HYSVUPBQPCRAJM-UHFFFAOYSA-L
Synonym 5-[4-(2-Carboxyethylcarbamoyl)phenylazo]salicylic Acid Disodium Salt
PubChem CID 53394935
IUPAC Name disodium;3-[[4-(2-carboxylatoethylcarbamoyl)phenyl]hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate;dihydrate
SMILES C1=CC(=CC=C1C(=O)NCCC(=O)[O-])NN=C2C=CC(=O)C(=C2)C(=O)[O-].O.O.[Na+].[Na+]

Specifications

Quantity 1 g
Formula Weight 401.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Balsalazide Disodium Salt Dihydrate