missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Balsalazide Disodium Salt Dihydrate 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 401.29 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Balsalazide Disodium Salt Dihydrate |
Safety and Handling
| TSCA | No |