missing translation for 'onlineSavingsMsg'
Learn More

Balsalazide disodium dihydrate, Thermo Scientific Chemicals

Catalog No. p-7214534
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465090010 1 g
AC465090050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465090010 Supplier Thermo Scientific Chemicals Supplier No. 465090010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150399-21-6
Molecular Formula C17H17N3Na2O8
Molecular Weight (g/mol) 437.32
InChI Key HYSVUPBQPCRAJM-PIFVRXTCSA-L
IUPAC Name disodium (3Z)-3-(2-{4-[(2-carboxylatoethyl)carbamoyl]phenyl}hydrazin-1-ylidene)-6-oxocyclohexa-1,4-diene-1-carboxylate dihydrate
SMILES O.O.[Na+].[Na+].[O-]C(=O)CCNC(=O)C1=CC=C(N\N=C2\C=CC(=O)C(=C2)C([O-])=O)C=C1

Specifications

Quantity 1 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: sparingly soluble in methanol and ethanol, practically insoluble in organic solvents
Formula Weight 437.32
Chemical Name or Material Balsalazide disodium dihydrate

RUO – Research Use Only