missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Balofloxacin Dihydrate 98.0+%, TCI America™
$130.48 - $397.05
Chemical Identifiers
| CAS | 151060-21-8 |
|---|---|
| Molecular Formula | C20H28FN3O6 |
| Molecular Weight (g/mol) | 425.457 |
| MDL Number | MFCD00864925 |
| InChI Key | WEGNYJXOCPJAPS-UHFFFAOYSA-N |
| Synonym | 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[3-(methylamino)-1-piperidyl]-4-oxoquinoline-3-carboxylic Acid |
| PubChem CID | 6918202 |
| IUPAC Name | 1-cyclopropyl-6-fluoro-8-methoxy-7-[3-(methylamino)piperidin-1-yl]-4-oxoquinoline-3-carboxylic acid;dihydrate |
| SMILES | CNC1CCCN(C1)C2=C(C=C3C(=C2OC)N(C=C(C3=O)C(=O)O)C4CC4)F.O.O |
Chemical Identifiers
| 151060-21-8 | |
| 425.457 | |
| WEGNYJXOCPJAPS-UHFFFAOYSA-N | |
| 6918202 | |
| CNC1CCCN(C1)C2=C(C=C3C(=C2OC)N(C=C(C3=O)C(=O)O)C4CC4)F.O.O |
| C20H28FN3O6 | |
| MFCD00864925 | |
| 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[3-(methylamino)-1-piperidyl]-4-oxoquinoline-3-carboxylic Acid | |
| 1-cyclopropyl-6-fluoro-8-methoxy-7-[3-(methylamino)piperidin-1-yl]-4-oxoquinoline-3-carboxylic acid;dihydrate |
Specifications
| 151060-21-8 | |
| White | |
| MFCD00864925 | |
| 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[3-(methylamino)-1-piperidyl]-4-oxoquinoline-3-carboxylic Acid | |
| CNC1CCCN(C1)C2=C(C=C3C(=C2OC)N(C=C(C3=O)C(=O)O)C4CC4)F.O.O | |
| 425.457 | |
| 389.43 | |
| Crystalline Powder |
| 135°C | |
| C20H28FN3O6 | |
| 1 g | |
| WEGNYJXOCPJAPS-UHFFFAOYSA-N | |
| 1-cyclopropyl-6-fluoro-8-methoxy-7-[3-(methylamino)piperidin-1-yl]-4-oxoquinoline-3-carboxylic acid;dihydrate | |
| 6918202 | |
| ≥98.0% (HPLC,T) | |
| Balofloxacin Dihydrate |
Safety and Handling
RTECSNumber : VB1993755
TSCA : No