missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Balloon - 12 Inch

Catalog No. S02946
Change view
Click to view available options
Class Size:
1 Student
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02946 1 Student
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02946 Supplier Eisco™ Supplier No. BALLOON12INCH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for educational experiments.

  • Made from high-quality, durable latex
  • Ideal for parties, events, and educational activities, offering vibrant colors and reliable performance
  • Suitable for helium or air inflation

Specifications

Grade Level High School
Grade K to 12
Material Latex
Quantity 1 Ea.
Size 12 in.
Class Size 1 Student
Product Type Balloons