missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Ballistics Car

Catalog No. S15119
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15119 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15119 Supplier Eisco™ Supplier No. PH0355APL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Ballistics Car demonstrates that the horizontal component of the velocity of a body thrown vertically upward from another body moving horizontally is independent of the vertical component of velocity.

  • Heavy metallic barrel mounted vertically on an aluminum car
  • Car features plastic wheels with ball bearings to minimize friction
  • Spring loaded piston arrangement with two compressions settings of spring for ejecting the ball to two different heights
  • Complete with steel ball and a lock pin with cord
  • Includes detailed instructions

Specifications

Product Type Ballistics Car
Grade 7 to 14
Material Aluminum (Car), Plastic (Wheels)
For Use With (Application) Velocity Demonstrations
Includes Car, Steel Ball, Lock Pin, Cord, and Instructions