missing translation for 'onlineSavingsMsg'
Learn More

Supertek™ Ball and Ring Apparatus
GREENER_CHOICE

Catalog No. S012333
Encompass
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S012333 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S012333 Supplier Supertek™ Supplier No. PH20022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An excellent device for demonstrating thermal expansion. When cool, the 25 mm brass ball passes easily through the matching brass ring.

  • Used for demonstrating thermal expansion.
  • When cool, the 25 mm brass ball passes easily through the matching brass ring.

Specifications

Grade 6+
Includes 1 ball, 1 ring
Class Size 1 Lab Group
Product Type Heat Conductometer