Learn More
Scientific Industries Inc BALANCE PRECISION 60 GRAM DFS Item
Supplier:  Scientific Industries Inc AD60
Precision Balance with 60g capacity, resolution of 1mg, rugged die-cast metal housing and highest quality electromagnetic load-cell. Loaded with 7 weighing functions all essential units of measure, equipped with RS232, USB, Clock, Backlit LCD Display.
Catalog No. 501139345
Only null left
Explore available promotions