Learn More
Scientific Industries Inc BALANCE PRECISION 520 GRAM DFS Item
Supplier:  Scientific Industries Inc AD520
Precision Balance with 520g capacity, resolution of 1mg, rugged die-cast metal housing and highest quality electromagnetic load-cell. Loaded with 7 weighing functions all essential units of measure, equipped with RS232, USB, Clock, Backlit LCD Display.
Catalog No. 501139349
Only null left
Explore available promotions